missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FDC Antibody (CNA.42), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP1-43118-0.1mg
This item is not returnable.
View return policy
Description
FDC Monoclonal specifically detects FDC in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
Specifications
| FDC | |
| Monoclonal | |
| 0.5 mg/mL | |
| Western Blot 1:100-1:2000, Flow Cytometry 1:10-1:1000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500 | |
| cardiomyopathy, dilated 1B (autosomal dominant), CMPD1, FDC | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| 1218 | |
| Store at 4C. Do not freeze. |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) | |
| CNA.42 | |
| Unconjugated | |
| Aqueous, may contain carrier protein/stabilizer. with 0.09% Sodium Azide | |
| CMD1B | |
| The immunogen for this antibody was follicular dendritic cells. | |
| 0.1 mg | |
| Primary | |
| Human | |
| IgM |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction