Learn More
Invitrogen™ FDFT1 Monoclonal Antibody (24GB5645)
GREENER_CHOICE
Mouse Monoclonal Antibody
Brand:  Invitrogen™ MA557398
 View more versions of this product
Product Code. 30276289
  • 564.00 € / 100µL
Description
FDFT1 Monoclonal Antibody for Western Blot
FDFT1 catalyzes the first step in the cholesterol biosynthetic pathway, the conversion of trans-farnesyldiphosphate to squalene. The loss of promoter activity and response to sterols for FDFT1 is localized to a 69-bp section positioned 131 bp 5-prime to the transcription start site. Sequence analysis of this region shows that it contains a sterol regulatory element-1 (SRE1) previously identified in other sterol regulated genes and 2 putative NF1 binding sites.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
FDFT1
Monoclonal
Unconjugated
FDFT1
DGPT; EGK_18680; ERG9; farnesyl diphosphate farnesyl transferase 1; farnesyl diphosphate farnesyltransferase; farnesyl-diphosphate farnesyltransferase; farnesyl-diphosphate farnesyltransferase 1; FDFT1; FPP:FPP farnesyltransferase; presqualene-di-diphosphate synthase; SQS; Squalene synthase; squalene synthetase; SS
Mouse
Affinity chromatography
RUO
2222
-20°C
Liquid
Flow Cytometry, Western Blot, Immunocytochemistry
24GB5645
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
P37268
FDFT1
Recombinant protein of human FDFT1 (aa 1-417).
100 μL
Primary
Human
Antibody
IgG2b