missing translation for 'onlineSavingsMsg'
Learn More

Febuxostat, MedChemExpress

Product Code. 16476426
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16476426 100 mg 100mg
16442157 50 mg 50mg
16426018 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16476426 Supplier Medchem Express Supplier No. HY14268
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Febuxostat (TEI 6720) is a potent, selective and non-purine xanthine oxidase (XO) inhibitor with a Ki value of 0.6 nM. Febuxostat has the potential for the research of hyperuricemia and gout.

Febuxostat (TEI 6720) is a potent, selective and non-purine xanthine oxidase (XO) inhibitor with a Ki value of 0.6 nM. Febuxostat has the potential for the research of hyperuricemia and gout.

Specifications

Chemical Name or Material Febuxostat
CAS 144060-53-7
Color White
Purity Grade Notes Research
Molecular Formula C16H16N2O3S
Quantity 100 mg
Synonym TEI 6720 TMX 67
Solubility Information DMSO : 50 mg/mL (158.04 mM; Need ultrasonic)
SMILES CC(COC1=CC=C(C=C1C#N)C2=NC(C)=C(S2)C(O)=O)C
Molecular Weight (g/mol) 316.37
Formula Weight 316.37
Percent Purity 99.74%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.