Learn More
Invitrogen™ FEN1 Recombinant Rabbit Monoclonal Antibody (24GB4835)
Rabbit Recombinant Monoclonal Antibody
Brand:  Invitrogen™ MA557403
 View more versions of this product
Product Code. 30275290
  • 198.00 € / 20µL
Description
FEN1 Recombinant Monoclonal Antibody for Western Blot, Flow
FEN-1 (Flap Endonuclease and Five prime Exonuclease) is a 42 a protein that functions as an endonuclease that cleaves branched DNA structures containing single-stranded 5' flaps or as an exonuclease on DNA nicks and gaps. FEN-1 endonuclease and exonuclease activities are stimulated by an association with proliferating cell nuclear antigen (PCNA) in the nucleus. FEN-1 is believed to have important roles in DNA replication and repair and is required in cell-free DNA replication systems.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
FEN1
Recombinant Monoclonal
Unconjugated
FEN1
AW538437; DNase IV; fen1; FEN-1; fen1 {ECO:0000255; FEN-1 {ECO:0000255; Flap endonuclease 1; flap endonuclease 1 {ECO:0000255; flap structure specific endonuclease 1; flap structure-specific endonuclease 1; flap structure-specific endonuclease 1 {ECO:0000255; HAMAP-Rule:MF_03140}; hFEN-1; Maturation factor 1; maturation factor-1; MF1; RAD2
Rabbit
Affinity chromatography
RUO
14156, 2237, 84490
-20°C
Liquid
Flow Cytometry, Western Blot, Immunocytochemistry
24GB4835
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
P39748, P39749, Q5XIP6
FEN1
A synthesized peptide derived from human FEN1 (aa 311-325).
20 μL
Primary
Human, Mouse, Rat