missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fenaminostrobin 100 μg/mL in Acetonitrile, Dr. Ehrenstorfer
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
Specifications
Specifications
| Chemical Name or Material | Fenaminostrobin |
| CAS | 366815-39-6 |
| Analyte or Component Names | Fenaminostrobin |
| Shipping Condition | +20°C |
| Molecular Formula | C21 H21 Cl2 N3 O3 |
| Concentration | 100 μg/mL |
| Certifications/Compliance | ISO 17034 |
| Synonym | Benzeneacetamide, 2-[[[(E)-[(2E)-3-(2,6-dichlorophenyl)-1-methyl-2-propenylidene]amino]oxy]methyl]-α-(methoxyimino)-N-methyl-, (αE)- (9CI), Diclofenoxystrobin, Fenaminostrobin, Fenaminstrobin, Xiwojunan, Benzeneacetamide, 2-[[[(E)-[(2E)-3-(2,6-dichlorophenyl)-1-methyl-2-propen-1-ylidene]amino]oxy]methyl]-α-(methoxyimino)-N-methyl-, (αE)- |
| InChI Formula | InChI=1S/C21H21Cl2N3O3/c1-14(11-12-17-18(22)9-6-10-19(17)23)25-29-13-15-7-4-5-8-16(15)20(26-28-3)21(27)24-2/h4-12H,13H2,1-3H3,(H,24,27)/b12-11+,25-14+,26-20+ |
| SMILES | CNC(=O)\C(=N\OC)\c1ccccc1CO\N=C(/C)\C=C\c2c(Cl)cccc2Cl |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction