missing translation for 'onlineSavingsMsg'
Learn More

Fenamiphos, Dr. Ehrenstorfer

Product Code. 30327768
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30327768 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30327768 Supplier Dr. Ehrenstorfer Supplier No. DREC13420000100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Fenamiphos
CAS 22224-92-6
Analyte or Component Names Fenamiphos
Shipping Condition +20°C
Molecular Formula C13 H22 N O3 P S
Certifications/Compliance ISO 17034
InChI Formula InChI=1S/C13H22NO3PS/c1-6-16-18(15,14-10(2)3)17-12-7-8-13(19-5)11(4)9-12/h7-10H,6H2,1-5H3,(H,14,15)
SMILES CCOP(=O)(NC(C)C)Oc1ccc(SC)c(C)c1
IUPAC Name N-[ethoxy-(3-methyl-4-methylsulfanylphenoxy)phosphoryl]propan-2-amine
Molecular Weight (g/mol) 303.36
Quantity 100 mg
Formula Weight 303.1058 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.