missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fenitrothion 10 μg/mL in Cyclohexane, Dr. Ehrenstorfer
Specifications
Specifications
| Chemical Name or Material | Fenitrothion |
| CAS | 122-14-5 |
| Analyte or Component Names | Fenitrothion |
| Shipping Condition | Room Temperature |
| Molecular Formula | C9 H12 N O5 P S |
| Concentration | 10 μg/mL |
| Certifications/Compliance | ISO 17034 |
| Synonym | Phosphorothioic acid, O,O-dimethyl O-(4-nitro-m-tolyl) ester (6CI,8CI), 8057HC, Accothion, Agria 1050, Agriya 1050, Agrothion, Arbogal, BAY 41831, Bayer 41831, Bayer S 5660, CL 47300, CP 47114, Dimethyl 4-nitro-m-tolyl phosphorothionate, EI 47300, ENT 25,715, Fenition, Fenitrothion, Folithion, Folithion EC 50, Insectigas F, MEP, MEP (pesticide), Metathion, Metathion E 50, Metathione, Metathionine E 50, Metation, Metation E 50, Methadion, Methylnitrophos, Mglawik F, Monsanto CP 47114, Nitrophos, Nuvanol, O,O-DiMe O-(3-methyl-4-nitrophenyl) thiophosphate, O,O-Dimethyl O-(3-methyl-4-nitrophenyl) phosphorothioate, O,O-Dimethyl O-(3-methyl-4-nitrophenyl) phosphorothioate, O,O-Dimethyl O-(3-methyl-4-nitrophenyl) phosphorothioate, O,O-Dimethyl O-(3-methyl-4-nitrophenyl) thiophosphate, O,O-Dimethyl O-(4-nitro-3-methylphenyl)thiophosphate, O,O-Dimethyl O-4-nitro-m-tolyl phosphorothioate, O,O-Dimethyl O-4-nitro-m-tolyl thiophosphate, OMS 43, Oleometathion, Oleosumifene, Ovadofos, Owadofos, Owadophos, Paluthion, Phenitrothion, S 112A, S 5660, ShaMingSong, Sumi oil, Sumifene, Sumigran, Sumithion, Sumithion 20F, Sumithion 20MC, Sumithion 50EC, Sumithion Super, Sumithion Super 1000EC, Super Sumithion, Tionfos 50 LE, Verthion |
| InChI Formula | InChI=1S/C9H12NO5PS/c1-7-6-8(4-5-9(7)10(11)12)15-16(17,13-2)14-3/h4-6H,1-3H3 |
| Solution Type | Single Solution |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?