missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Fenofibrate, 98%

Product Code. 15790909 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15790909 5 g 5g
15700919 25 g 25g
15710919 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15790909 Supplier Thermo Scientific Chemicals Supplier No. 461780050
ChemsChrom Mega Deals
Use promo code 27993
View all promotions
 Restricted Item
Estimated Shipment: 25-11-2026
Add to basket
Edge

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 49562-28-9 | C20H21ClO4 | 360.83 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

Molecular Formula C20H21ClO4
Molecular Weight (g/mol) 360.83
InChI Key YMTINGFKWWXKFG-UHFFFAOYSA-N
PubChem CID 3339
ChEBI CHEBI:5001

Specifications

Melting Point 79°C to 82°C
Assay Percent Range 97.5% min. (HPLC)
Quantity 5 g
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in ether,soluble in acetone,,soluble in ethyl acetate,soluble in acetonitrile,soluble in methanol,soluble in ethanol
Formula Weight 360.83
Percent Purity 98%
Chemical Name or Material Fenofibrate
compliance-icons
Product Identifier
  • Fenofibrate, 98%
Signal Word
  • Warning
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 4
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H373-May cause damage to organs.
  • H413-May cause long lasting harmful effects to aquatic life.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
  • P314-Get medical advice/attention if you feel unwell.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: Fenofibrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.