missing translation for 'onlineSavingsMsg'
Learn More

Fenoprofen Calcium hydrate, MedChemExpress

Product Code. 16412129
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16412129 1 g 1g
16400868 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16412129 Supplier Medchem Express Supplier No. HYB0288B
 Restricted Item
Estimated Shipment: 10-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Fenoprofen Calcium hydrate is a nonsteroidal, anti-inflammatory antiarthritic agent.

Fenoprofen Calcium hydrate is a nonsteroidal, anti-inflammatory antiarthritic agent.

Specifications

Chemical Name or Material Fenoprofen Calcium hydrate
CAS 71720-56-4
Color White
Purity Grade Notes Research
Molecular Formula C15H15Ca0.5O4
Quantity 1 g
Synonym Fenoprofen calcium salt dihydrate
Solubility Information DMSO : ≥ 100 mg/mL (358.01 mM) ∣H2O : 1 mg/mL (3.58 mM; Need ultrasonic)
SMILES CC(C([O-])=O)C1=CC(OC2=CC=CC=C2)=CC=C1.O.[0.5Ca2+]
Molecular Weight (g/mol) 279.32
Formula Weight 279.32
Percent Purity 99.93%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.