missing translation for 'onlineSavingsMsg'
Learn More

Fenoprofen Calcium Salt Dihydrate, TRC

Product Code. 30012298
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30012298 5 g 5g
30005805 25 g 25g
30007032 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30012298 Supplier Toronto Research Chemicals Supplier No. TRCF2488255G
 Restricted Item
Estimated Shipment: 14-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 71720-56-4
Molecular Formula 2 C15 H13 O3 . Ca . 2 H2 O
Molecular Weight (g/mol) 558.63
Synonym Fenoprofen Calcium Dihydrate, Benzeneacetic acid, α-methyl-3-phenoxy-, calcium salt, hydrate (2:1:2), Benzeneacetic acid, α-methyl-3-phenoxy-, calcium salt, dihydrate (9CI), Fenoprofen calcium salt dihydrate
IUPAC Name calcium;2-(3-phenoxyphenyl)propanoate;dihydrate
SMILES O.O.[Ca+2].CC(C(=O)[O-])c1cccc(Oc2ccccc2)c1.CC(C(=O)[O-])c3cccc(Oc4ccccc4)c3

Specifications

Quantity 5 g
InChI Formula InChI=1S/2C15H14O3.Ca.2H2O/c2*1-11(15(16)17)12-6-5-9-14(10-12)18-13-7-3-2-4-8-13;;;/h2*2-11H,1H3,(H,16,17);;2*1H2/q;;+2;;/p-2
Formula Weight 558.1567
Chemical Name or Material Fenoprofen calcium dihydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.