missing translation for 'onlineSavingsMsg'
Learn More

Fenoprofen Sodium Salt Dihydrate, TRC

Product Code. 30132457
Change view
Click to view available options
Quantity:
500 mg
5 g
Unit Size:
500mg
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30132457 5 g 5g
30012365 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30132457 Supplier Toronto Research Chemicals Supplier No. TRCF2488305G
 Restricted Item
Estimated Shipment: 14-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 66424-46-2
Molecular Formula C15 H13 O3 . Na . 2 H2 O
Molecular Weight (g/mol) 300.28
Synonym Benzeneacetic acid, α-methyl-3-phenoxy-, sodium salt, hydrate (1:1:2), Benzeneacetic acid, α-methyl-3-phenoxy-, sodium salt, dihydrate (9CI), Benzeneacetic acid, α-methyl-3-phenoxy-, sodium salt, dihydrate, (±)-, Fenoprofen sodium salt dihydrate
IUPAC Name sodium;2-(3-phenoxyphenyl)propanoate;dihydrate
SMILES O.O.[Na+].CC(C(=O)[O-])c1cccc(Oc2ccccc2)c1

Specifications

Quantity 5 g
InChI Formula InChI=1S/C15H14O3.Na.2H2O/c1-11(15(16)17)12-6-5-9-14(10-12)18-13-7-3-2-4-8-13;;;/h2-11H,1H3,(H,16,17);;2*1H2/q;+1;;/p-1
Formula Weight 300.0974
Chemical Name or Material Fenoprofen Sodium Dihydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.