missing translation for 'onlineSavingsMsg'
Learn More

Fenothiocarb, Dr. Ehrenstorfer

Product Code. 30314138
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30314138 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30314138 Supplier Dr. Ehrenstorfer Supplier No. DREC13497500
 Restricted Item
Estimated Shipment: 30-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Fenothiocarb
CAS 62850-32-2
Analyte or Component Names Fenothiocarb
Shipping Condition Room Temperature
Molecular Formula C13 H19 N O2 S
Certifications/Compliance ISO 17034
Synonym S-(4-Phenoxybutyl) Dimethylcarbamothioate, S-(4-Phenoxybutyl) Ester N,N-Dimethyl-carbamothioic Acid, Panocon, Phenothiocarb;
InChI Formula InChI=1S/C13H19NO2S/c1-14(2)13(15)17-11-7-6-10-16-12-8-4-3-5-9-12/h3-5,8-9H,6-7,10-11H2,1-2H3
Solution Type Neat
SMILES CN(C)C(=O)SCCCCOc1ccccc1
IUPAC Name S-(4-phenoxybutyl) N,N-dimethylcarbamothioate
Industry Type Environmental Testing
Molecular Weight (g/mol) 253.36
Quantity 100 mg
Formula Weight 253.1136 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.