missing translation for 'onlineSavingsMsg'
Learn More

Fensulfothion 1000 μg/mL in Toluene, Dr. Ehrenstorfer

Product Code. 30325299
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30325299 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30325299 Supplier Dr. Ehrenstorfer Supplier No. DREA13570000TO1000
 Restricted Item
Estimated Shipment: 07-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Fensulfothion
CAS 115-90-2
Analyte or Component Names Fensulfothion
Shipping Condition +20°C
Molecular Formula C11 H17 O4 P S2
Concentration 1000 μg/mL
Certifications/Compliance ISO 17034
Synonym Phosphorothioic acid, O,O-diethyl O-[4-(methylsulfinyl)phenyl] ester, Phosphorothioic acid, O,O-diethyl O-[p-(methylsulfinyl)phenyl] ester (6CI,7CI,8CI), Phenol, p-(methylsulfinyl)-, O-ester with O,O-di-Et phosphorothioate (6CI), Agricur, B 25141, BAY 25141, Chemagro 25141, DMSP, Daconit, Dasanit, ENT 24,945, Fensulfothion, O,O-Diethyl O-(4-methylsulfinylphenyl) monothiophosphate, O,O-Diethyl O-[4-(methylsulfinyl)phenyl] phosphorothioate, O,O-Diethyl O-[p-(methylsulfinyl)phenyl] phosphorothioate, S 767, Terracur P, VUAgt 108, VUAgt 96
InChI Formula InChI=1S/C11H17O4PS2/c1-4-13-16(17,14-5-2)15-10-6-8-11(9-7-10)18(3)12/h6-9H,4-5H2,1-3H3
SMILES CCOP(=S)(OCC)Oc1ccc(cc1)S(=O)C
IUPAC Name diethoxy-(4-methylsulfinylphenoxy)-sulfanylidene-λ^{5}-phosphane
Solvent or Matrix Toluene
Molecular Weight (g/mol) 308.35
Quantity 1 mL
Formula Weight 308.0306 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.