missing translation for 'onlineSavingsMsg'
Learn More

Fenthion 1000 μg/mL in Toluene, Dr. Ehrenstorfer

Product Code. 30319932
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30319932 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30319932 Supplier Dr. Ehrenstorfer Supplier No. DREA13580000TO1000

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Fenthion
CAS 55-38-9
Analyte or Component Names Fenthion
Shipping Condition +20°C
Molecular Formula C10 H15 O3 P S2
Concentration 1000 μg/mL
Certifications/Compliance ISO 17034
Synonym Fenthion
InChI Formula InChI=1S/C10H15O3PS2/c1-8-7-9(5-6-10(8)16-4)13-14(15,11-2)12-3/h5-7H,1-4H3
SMILES COP(=S)(OC)Oc1ccc(SC)c(C)c1
IUPAC Name dimethoxy-(3-methyl-4-methylsulfanylphenoxy)-sulfanylidene-λ^{5}-phosphane
Solvent or Matrix Toluene
Molecular Weight (g/mol) 278.33
Quantity 1 mL
Formula Weight 278.02 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.