missing translation for 'onlineSavingsMsg'
Learn More

Fenticonazole Nitrate, MedChemExpress

Product Code. 16464337
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16464337 50 mg 50mg
16428458 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16464337 Supplier Medchem Express Supplier No. HYB0359
 Restricted Item
Estimated Shipment: 03-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Fenticonazole Nitrate is an antifungal imidazole ring derivative. Fenticonazole Nitrate operates via hindering ergosterol integration, and sequentially destructing the cytoplasmatic outer membrane. Fenticonazole Nitrate is effective against Gram-positive bacteria, mycoses, and vaginal candidiasis.

Fenticonazole Nitrate is an antifungal imidazole ring derivative. Fenticonazole Nitrate operates via hindering ergosterol integration, and sequentially destructing the cytoplasmatic outer membrane. Fenticonazole Nitrate is effective against Gram-positive bacteria, mycoses, and vaginal candidiasis.

Specifications

Chemical Name or Material Fenticonazole Nitrate
Physical Form Solid
CAS 73151-29-8
Color Off-White
Quantity 50 mg
Purity Grade Notes Research
Molecular Formula C24H21Cl2N3O4S
For Use With (Application) COVID-19-immunoregulation
Synonym REC 15-1476
Solubility Information DMSO : ≥ 100 mg/mL (192.90 mM)
SMILES [O-][N+](O)=O.ClC1=CC=C(C(OCC2=CC=C(SC3=CC=CC=C3)C=C2)CN4C=CN=C4)C(Cl)=C1
Molecular Weight (g/mol) 518.41
Formula Weight 518.41
Percent Purity 99.44%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.