missing translation for 'onlineSavingsMsg'
Learn More

Ferulic Acid, 99.4%, MP Biomedicals™

Product Code. 11468050 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11468050 100 g 100g
11448050 5 g 5g
11458050 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11468050 Supplier MP Biomedicals Supplier No. 0210168580
 Restricted Item
Estimated Shipment: 10-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 1135-24-6
Molecular Formula C10H10O4
Molecular Weight (g/mol) 194.186
InChI Key KSEBMYQBYZTDHS-HWKANZROSA-N
Synonym ferulic acid, trans-ferulic acid, 4-hydroxy-3-methoxycinnamic acid, trans-4-hydroxy-3-methoxycinnamic acid, 3-4-hydroxy-3-methoxyphenyl acrylic acid, e-ferulic acid, coniferic acid, ferulate, ferulic acid, trans, 2-propenoic acid, 3-4-hydroxy-3-methoxyphenyl
PubChem CID 445858
ChEBI CHEBI:17620
IUPAC Name (E)-3-(4-hydroxy-3-methoxyphenyl)prop-2-enoic acid
SMILES COC1=C(C=CC(=C1)C=CC(=O)O)O

Specifications

Assay Percent Range 99.4%
Quantity 100 g
Formula Weight 194.19
Physical Form Crystalline
Chemical Name or Material Ferulic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.