missing translation for 'onlineSavingsMsg'
Learn More

Ferulic Acid, TRC

Product Code. 30007272
Change view
Click to view available options
Quantity:
50 mg
100 mg
1 g
Unit Size:
100mg
1g
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30007272 1 g 1g
17909074 100 mg 100mg
17990065 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30007272 Supplier Toronto Research Chemicals Supplier No. TRCF3089001G
 Restricted Item
Estimated Shipment: 03-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1135-24-6
Molecular Formula C10 H10 O4
Molecular Weight (g/mol) 194.18
Synonym Cinnamic acid, 4-hydroxy-3-methoxy- (7CI,8CI), 3-(4-Hydroxy-3-methoxyphenyl)-2-propenoic acid, 3-(4-Hydroxy-3-methoxyphenyl)acrylic acid, 3-Methoxy-4-hydroxycinnamic acid, 4-Hydroxy-3-methoxycinnamic acid, 4'-Hydroxy-3'-methoxycinnamic acid, Coniferic acid, Ferulaic acid, Ferulic acid, NSC 2821, NSC 51986, NSC 674320
IUPAC Name 3-(4-hydroxy-3-methoxyphenyl)prop-2-enoic acid
SMILES COc1cc(C=CC(=O)O)ccc1O

Specifications

Quantity 1 g
InChI Formula InChI=1S/C10H10O4/c1-14-9-6-7(2-4-8(9)11)3-5-10(12)13/h2-6,11H,1H3,(H,12,13)
Formula Weight 194.0579
Percent Purity >95%
Chemical Name or Material Ferulic Acid
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.