Learn More
Chemsplash FFP2 Cup Mask with Valve
Product Code. 30058359
Change view
Click to view available options
Aerosol Type:
N - Not Resistant to Oil
Unit Size:
Pack of 200
Product Code. Aerosol Type Unit Size
30058359 N - Not Resistant to Oil Pack of 200
1 options
Product Code. 30058359 Supplier Chemsplash Supplier No. 3672
  • ONLINE EXCLUSIVE PRICE
    129.93 €
    120.18 € / Pack of 200
    Save 9.75 €
    7% Off
Estimated Shipment: 18-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
The FFP2 valved cup mask has a high filtering efficiency. It has woven elasticated adjustable head straps.
Chemsplash 3672 FFP2 Valved Cup Mask
  • High Filtration Capacity
  • Respirator with Yellow Color Exhalation Valve
  • Exhalation Valve Reduces Hot Air Buildup
  • Woven Elasticated Adjustable Head Straps
  • Adjustable Nose Clip with Soft Inner Nose Foam
  • Protects Against Low-Toxicity Solid and Liquid Particles
  • No Vision Impairment and Problem-Free Speech
  • Latex and Graphene Free
Description: The Chemsplash 3672 is an FFP2 valved cup mask, colour-coded yellow for ease of use. The fabric material of the mask has a high filtering efficiency. It features woven elasticated adjustable head straps and a soft padded band that rests inside, adjustable for an excellent fit on the nose. This mask provides reliable protection against low-toxicity solid and liquid particles while ensuring no impairment to vision and problem-free speech. It is latex and graphene-free for enhanced safety and comfort.
Specifications
Aerosol Type N - Not Resistant to Oil
Color White
Design Type Cup/Molded
Disposable Single-use
Filter Type FFP2
Material Polypropylene
Product Type General Purpose Particulate Respirator
Size One Size
With Exhalation Valve Yes
Latex Free Yes
Sterility Non-sterile