Learn More
Chemsplash FFP2 Fold Flat into Cup Mask
Product Code. 30058471
Change view
Click to view available options
Aerosol Type:
N - Not Resistant to Oil
Unit Size:
Pack of 200
Product Code. Aerosol Type Unit Size
30058471 N - Not Resistant to Oil Pack of 200
1 options
Product Code. 30058471 Supplier Chemsplash Supplier No. 3508
  • ONLINE EXCLUSIVE PRICE
    187.44 €
    173.64 € / Pack of 200
    Save 13.80 €
    7% Off
Estimated Shipment: 18-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
The FFP2 valved fold flat into cup mask has super fold flat design, 3D Structure and Large Filtration Area.
Chemsplash 3508 FFP2 Valved Fold Flat into Cup Mask
  • High Filtration Capacity
  • Respirator with Yellow Color Exhalation Valve
  • Exhalation Valve Reduces Hot Air Buildup
  • Adjustable Nose Clip with Padded Comfort on Inside
  • Woven Elasticated Adjustable Head Straps
  • Super Fold Flat Design, 3D Structure and Large Filtration Area
  • Pass the Optional Dolomite (D) Clogging Test
  • Packed Individually for Hygiene
  • Protects Against Low-Toxicity Solid and Liquid Particles
  • Latex and Graphene Free
Description: The Chemsplash 3508 is a FFP2 valved fold flat into cup mask, featuring a super fold flat design, 3D structure, and a large filtration area. It is colour-coded yellow for ease of use. The mask fabric has high filtering efficiency and passes the optional Dolomite Clogging Test. An adjustable nose clip with padded comfort ensures an excellent fit, while the woven elasticated adjustable head straps offer secure and comfortable wear. This mask protects against low-toxicity solid and liquid particles and is latex and graphene-free. It is packed individually for hygiene.
Specifications
Aerosol Type N - Not Resistant to Oil
Color White
Design Type Flat-fold
Disposable Single-use
Filter Type FFP2
Material Polypropylene
Product Type General Purpose Particulate Respirator
Size One Size
With Exhalation Valve Yes
Latex Free Yes
Sterility Non-sterile