Learn More
Chemsplash FFP2 Fold Flat Mask with Valve
Product Code. 30058348
Change view
Click to view available options
Aerosol Type:
N - Not Resistant to Oil
Unit Size:
Pack of 200
Product Code. Aerosol Type Unit Size
30058348 N - Not Resistant to Oil Pack of 200
1 options
Product Code. 30058348 Supplier Chemsplash Supplier No. 3673
  • ONLINE EXCLUSIVE PRICE
    124.16 €
    115.82 € / Pack of 200
    Save 8.34 €
    6% Off
View all promotions
Estimated Shipment: 18-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
The FFP2 fold flat valved mask has an exhalation valve which reduces hot air buildup.
Chemsplash 3673 FFP2 Fold Flat Valved Mask
  • High Filtration Capacity
  • Respirator with Yellow Color Exhalation Valve
  • Exhalation Valve Reduces Hot Air Buildup
  • Woven Elasticated Adjustable Head Straps
  • Adjustable Nose Clip with Soft Inner Nose Foam
  • Folded to Pocket Size and Packed Individually for Hygiene
  • Protects Against Low-Toxicity Solid and Liquid Particles
  • No Vision Impairment and Problem-Free Speech
  • Latex and Graphene Free
Description: The Chemsplash 3673 is an FFP2 fold-flat valved mask that comes with an exhalation valve to reduce hot air buildup. Color-coded yellow for easy identification, this mask is made of high filtration fabric, protecting against low-toxicity solid and liquid particles. A soft padded band rests inside, adjustable to ensure an excellent fit on the nose. The mask is folded to pocket size and individually packed for hygiene. It offers great comfort and ensures no vision impairment or speech difficulty. Additionally, it is latex and graphene-free for safer use.
Specifications
Aerosol Type N - Not Resistant to Oil
Color White
Design Type Flat-fold
Disposable Single-use
Filter Type FFP2
Material Polypropylene
Product Type General Purpose Particulate Respirator
Size One Size
With Exhalation Valve Yes
Latex Free Yes
Sterility Non-sterile