missing translation for 'onlineSavingsMsg'
Learn More

New Source™ FFP3 Disposable Respirator Face Mask

Product Code. p-9090002
Click to view available options
With Exhalation Valve:
No
Yes
Unit Size:
Pack of 10
Pack of 20
This item is not returnable. View return policy

Product Code. 16679272

Brand: New Source™ HY9330

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

FFP3 Disposable respirator to protect against inhalation of droplets and particles. Available with or without exhalation valve.

  • Polypropylene outer layers
  • Fold flat
  • 16689272 has exhalation valve for increased comfort
  • 16679272 is without valve
  • Compatible with goggles/glasses and reduces fogging due to contoured design
  • Adjustable soft-foam nosepiece ensures custom fit to increase wearer's comfort
  • Seal avoids open layer around the edges

Specifications

Certifications/Compliance CE Approved, EN 149:2001+A1:2009 FFP3 NR D
Color White
Design Type Flat-fold
Disposable Single-use
Material Anti-Bacterial Cotton, Non-Woven Fabric
Product Type Particulate Respirator with Double Head Bands
Size One Size
With Exhalation Valve No
Filter Efficiency FFP3
For Use With (Application) Protection Against Dusts and Mists in a Wide Variety of Industrial Applications and Other Hazardous Environments
Latex Free Yes
Sterility Non-sterile
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.