missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FGF-9 Antibody (FG9-77), Alexa Fluor™ 488, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB600-623AF488
This item is not returnable.
View return policy
Description
FGF-9 Monoclonal specifically detects FGF-9 in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunohistochemistry.
Specifications
| FGF-9 | |
| Monoclonal | |
| Alexa Fluor 488 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| FGF9 | |
| Recombinant mouse FGF-9 | |
| 0.1 mL | |
| Angiogenesis, Biologically Active Proteins, Cancer, Cell Cycle and Replication | |
| 2254 | |
| Human, Mouse | |
| Purified |
| Western Blot, ELISA, Immunohistochemistry | |
| FG9-77 | |
| Western Blot, ELISA, Immunohistochemistry | |
| FGF-9, Fibroblast growth factor 9, fibroblast growth factor 9 (glia-activating factor), GAF, glia-activating factor, HBFG-9, HBGF-9, Heparin-binding growth factor 9, MGC119914, MGC119915, SYNS3 | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Monoclonal Anti-FGF9 reacts specifically with recombinant FGF9 (fibroblast growth factor 9). It does not cross-react with human FGF Acidic and FGF Basic. | |
| Store at 4C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction