Learn More
Novus Biologicals™ FGF basic/FGF2/bFGF Antibody (HuGAL-F2) - Humanized, Novus Biologicals™
Human Monoclonal Antibody
Brand:  Novus Biologicals™ NBP3-28540-1mg
 View more versions of this product
Product Code. 30046918
  • 920.00 € / 1mg
Description
FGF basic/FGF2/bFGF Monoclonal antibody specifically detects FGF basic/FGF2/bFGF in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
TRUSTED_SUSTAINABILITY
Specifications
Specifications
FGF basic/FGF2/bFGF
Monoclonal
Unconjugated
P09038
Human
Protein A purified
RUO
Primary
Reconstitute with sterile, distilled water to a final concentration of 1 mg/mL. Gently shake to solubilize completely. Do not vortex.
Store at -20°C in powder form. Store at -80°C once reconstituted.
IgG2
Flow Cytometry, ELISA, Functional Assay
HuGAL-F2
Lyophilized from 25mM histidine, 8% sucrose, 0.01% Tween80 (pH6.2)
Basic fibroblast growth factor, basic fibroblast growth factor bFGF, bFGF, FGFBprostatropin, fibroblast growth factor 2 (basic), HBGF-2, heparin-binding growth factor 2
FGF2
1 mg
Angiogenesis, Biologically Active Proteins, Cancer
2247
Human
Purified