missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FGFR2 Rabbit anti-Human, Clone: 16, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-89673-50UL
This item is not returnable.
View return policy
Description
FGFR2 Monoclonal specifically detects FGFR2 in Human samples. It is validated for ELISA.
Specifications
| FGFR2 | |
| Monoclonal | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| ELISA | |
| 16 | |
| ELISA 1:5000-1:10000 | |
| BBDS, BEK, BFR-1, CD332, CD332 antigen, CEK3, CFD1, craniofacial dysostosis 1, EC 2.7.10, EC 2.7.10.1, ECT1, fibroblast growth factor receptor 2, FLJ98662, Jackson-Weiss syndrome, JWS, Keratinocyte growth factor receptorreceptor like 14, KGFR, K-sam, KSAM, soluble FGFR4 variant 4, TK14, TK25 | |
| Recombinant Human FGFR2 protein | |
| 50 μg | |
| Angiogenesis, Protein Kinase | |
| 2263 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction