missing translation for 'onlineSavingsMsg'
Learn More

FH 535, Tocris Bioscience™

Product Code. 18782694
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18782694 10 mg 10mg
18772514 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18782694 Supplier Tocris Bioscience Supplier No. 4344/10

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Inhibitor of Wnt/β-catenin signaling

Inhibitor of Wnt/β-catenin signaling and dual antagonist of PPARγ/δ activity. Suppresses β-catenin/Tcf-mediated transcription; inhibits β-catenin and GRIP1 recruitment to PPARγ and δ. Exhibits antiproliferative effects in transformed colon, lung and liver cancer cell lines.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 108409-83-2
Molecular Formula C13H10Cl2N2O4S
Molecular Weight (g/mol) 361.19
MDL Number MFCD01212888
InChI Key AXNUEXXEQGQWPA-UHFFFAOYSA-N
Synonym 2,5-dichloro-n-2-methyl-4-nitrophenyl benzenesulfonamide, beta-catenin/tcf inhibitor, fh535, n-2-methyl-4-nitrophenyl-2,5-dichlorobenzenesulfonamide, cbmicro_011295, fh hplc
PubChem CID 3463933
IUPAC Name 2,5-dichloro-N-(2-methyl-4-nitrophenyl)benzene-1-sulfonamide
SMILES CC1=C(NS(=O)(=O)C2=C(Cl)C=CC(Cl)=C2)C=CC(=C1)[N+]([O-])=O

Specifications

Quantity 10 mg
Formula Weight 361.2
Percent Purity >99%
Chemical Name or Material FH 535
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.