missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FH Rabbit, anti-Human, CoraLite Plus 488, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
145.00 € - 450.00 €
Specifications
| Antigen | FH |
|---|---|
| Concentration | 1000 μg/mL |
| Applications | Immunofluorescence |
| Conjugate | CoraLite Plus 488 |
| Form | Liquid |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18067025
|
Proteintech
CL488-11375-20UL |
20 μL |
145.00 €
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18095475
|
Proteintech
CL488-11375-100UL |
100 μL |
450.00 €
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The FH antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human FH. This antibody recognizes human, mouse, rat antigen. The FH antibody has been validated for the following applications: IF analysis. This antibody is coralite™ plus 488.
Specifications
| FH | |
| Immunofluorescence | |
| Liquid | |
| Human, Mouse, Rat | |
| FH, Fumarase, fumarate hydratase, HLRCC, LRCC, MCL, MCUL1 | |
| Primary |
| 1000 μg/mL | |
| CoraLite Plus 488 | |
| Rabbit | |
| 2271 | |
| IgG | |
| Store at -20°C. Avoid exposure to light. Stable for one year after shipment. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title