missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FH Rabbit Recombinant Antibody, Proteintech
Rabbit Recombinant Antibody
129.00 € - 399.00 €
Specifications
| Antigen | FH |
|---|---|
| Clone | 3K16 |
| Concentration | 500 μg/mL |
| Applications | Western Blot, Immunoprecipitation, ELISA |
| Classification | Recombinant Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30237749
|
Proteintech
82508-1-RR-20UL |
20 μL |
149.00 € 129.00 € / 20µL Save 20.00 € 13% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30234286
|
Proteintech
82508-1-RR-100UL |
100 μL |
439.00 € 399.00 € / 100µL Save 40.00 € 9% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The FH antibody from Proteintech is a rabbit recombinant antibody generated with recombinant protein of human FH. This antibody recognizes human, rat, mouse antigen. The FH antibody has been validated for the following applications: WB, IP, ELISA analysis.Specifications
| FH | |
| 500 μg/mL | |
| Recombinant Monoclonal | |
| Liquid | |
| RUO | |
| 2271 | |
| Recombinant Protein | |
| Primary | |
| Store at -20°C. Stable for one year after shipment. |
| 3K16 | |
| Western Blot, Immunoprecipitation, ELISA | |
| Unconjugated | |
| Rabbit | |
| Human, Rat, Mouse | |
| HLRCC; LRCC; MCL; MCUL1 | |
| IgG | |
| Protein A purification | |
| Recombinant Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title