missing translation for 'onlineSavingsMsg'
Learn More

FH535, MedChemExpress

Product Code. 16466768
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16466768 10 mg 10mg
16452847 50 mg 50mg
16436896 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16466768 Supplier Medchem Express Supplier No. HY15721
 Restricted Item
Estimated Shipment: 11-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress FH535 is an inhibitor of Wnt/β-catenin and PPAR, with anti-tumor activities.

FH535 is an inhibitor of Wnt/β-catenin and PPAR, with anti-tumor activities.

Specifications

Chemical Name or Material FH535
CAS 108409-83-2
Color Yellow
Purity Grade Notes Research
Molecular Formula C13H10Cl2N2O4S
Quantity 10 mg
Solubility Information DMSO : 33.33 mg/mL (92.28 mM; Need ultrasonic)
SMILES O=S(C1=CC(Cl)=CC=C1Cl)(NC2=CC=C([N+]([O-])=O)C=C2C)=O
Molecular Weight (g/mol) 361.2
Formula Weight 361.2
Percent Purity 99.87%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.