missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FHL3 Rabbit anti-Human, Mouse, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
197.00 € - 470.00 €
Specifications
| Antigen | FHL3 |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Form | Purified |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18025306
|
Novus Biologicals
NBP3-04622-20UL |
20 μL |
197.00 €
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18322656
|
Novus Biologicals
NBP3-04622-100UL |
100 μL |
470.00 €
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
FHL3 Polyclonal specifically detects FHL3 in Human, Mouse samples. It is validated for Western Blot.Specifications
| FHL3 | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS with 50% glycerol, pH7.3. | |
| 2275 | |
| IgG | |
| Affinity purified |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse | |
| FHL-3, four and a half LIM domains 3, four and a half LIM domains protein 3, LIM-only protein FHL3, MGC19547, MGC8696, Skeletal muscle LIM-protein 2, SLIM-2, SLIM2MGC23614 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-280 of human FHL3 (NP_004459.2). | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title