missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Fibrin Degradation Product E Native Protein

Product Code. 18651489 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18651489 0.1 mg 0.1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18651489 Supplier Novus Biologicals™ Supplier No. NBP262457
  • ONLINE EXCLUSIVE PRICE
    881.00 €
    832.65 € / 0.1mg
    Save 48.35 €
    5% Off
 Restricted Item
Estimated Shipment: 13-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Native Protein

TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) ELISA
Formulation Glycine buffered saline
Molecular Weight (g/mol) MolecularWeight-theroretical: 50 kDa
Quantity 0.1 mg
Storage Requirements Store at 4°C short term. Store at -20°C long term. Avoid freeze/thaw cycles.
Reconstitution Reconstitute with 1.0 mL distilled water. Care should be taken during reconstitution as the protein may appear as a film (at the bottom of the vial. Gently mix vial after reconstitution.)
Research Category Angiogenesis, Cancer, Cell Cycle and Replication
Protein Fibrin Degradation Product E
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.