missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Fibroblasts/Epithelial Cells Monoclonal Antibody (D7-FIB)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA528455
This item is not returnable.
View return policy
Description
This antibody recognizes approximately a 112kDa molecule expressed on the cell surface of human fibroblasts. The antigen is not expressed by peripheral blood cells, and is found at low levels on a minority of melanoma cell lines.
Fibroblasts/Epithelial Cells antibody recognizes an approximately 112 kDa molecule expressed on the cell surface of human fibroblasts. The antigen is not expressed by peripheral blood cells, and is found at low levels on a minority of melanoma cell lines. Studies upon the antigen have shown it to be sensitive to SDS, but resistant to trypsin, tunicamycin and collagenase.
Specifications
| Fibroblasts/Epithelial Cells | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 0.09% sodium azide | |
| Mouse | |
| Protein A | |
| RUO | |
| Human | |
| Antibody | |
| IgG2a |
| Flow Cytometry, Immunohistochemistry (Frozen), Immunoprecipitation | |
| D7-FIB | |
| Unconjugated | |
| fibro-blasts | |
| Human foreskin fibroblasts. | |
| 200 μg | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction