missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Fibromodulin Monoclonal Antibody (549302)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA561606
This item is not returnable.
View return policy
Description
Reconstitute at 0.5 mg/mL in sterile PBS.
Fibromodulin is a member of a family of small interstitial proteoglycans, containing a central region composed of leucine-rich repeats with 4 keratan sulfate chains flanked by disulfide-bonded terminal domains. It may participate in the assembly of the extracellular matrix as it interacts with type I and type II collagen fibrils and inhibits fibrillogenesis in vitro. It may also regulate TGF-beta activities by sequestering TGF-beta into the extracellular matrix.
Specifications
| Fibromodulin | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with trehalose and no preservative | |
| Q06828 | |
| FMOD | |
| Mouse myeloma cell line NS0-derived recombinant human Fibromodulin/FMOD, Asp75-Ile376. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b |
| Western Blot | |
| 549302 | |
| Unconjugated | |
| FMOD | |
| AI131919; AU041740; Collagen-binding 59 kDa protein; fibromodulin; FM; FMOD; Keratan sulfate proteoglycan fibromodulin; KSPG fibromodulin; SLRR2E | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 2331 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction