missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fibronectin Antibody (A17), DyLight 405, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-23524V
This item is not returnable.
View return policy
Description
Fibronectin Monoclonal antibody specifically detects Fibronectin in Human, Bovine, Chicken samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)
Specifications
| Fibronectin | |
| Monoclonal | |
| DyLight 405 | |
| CIG, ED-B, fibronectin 1, FINC, FN, FN1, FNZ, GFND, GFND2, LETS, MSF, SMDCF | |
| Lysed bovine corneal endothelial cells and extracellular matrix | |
| 100 μg | |
| Cancer, Cellular Markers, Extracellular Matrix, Mesenchymal Stem Cell Markers, Stem Cell Markers | |
| 2335 | |
| Store at 4°C in the dark. | |
| IgG1 |
| Western Blot, ELISA, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
| A17 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human, Bovine, Chicken | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction