Learn More
Invitrogen™ Fibronectin Monoclonal Antibody (A17)

Description
CSI 005-17-02 is highly specific for fibronectin. There is no evidence for cross-reactivity with other connective tissue proteins (vitronectin, elastin, collagen, laminin). CSI 005-17-02 cross-reacts with human and chicken fibronectin. Other species have not been tested. CSI 005-17-02 has been successfully used in Western blottin, ELISA, Immunohistochemistry and Immunoprecipitation procedures. The antibody inhibits integrin-mediated cell adhesion to the cell binding domain of fibronectin. It can be used to probe fibronectin conformation. Strong reaction is seen in ELISA with thrombospondin directly coated onto the microtiter well. The specificity of the CSI 005-17-02 epitope is located in the 120 kDa cell binding fragment. NOTE: Concentration is lot-dependent and can vary from 0.85-1.15 mg/mL.
Specifications
Specifications
| Antigen | Fibronectin |
| Applications | ELISA, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot |
| Classification | Monoclonal |
| Clone | A17 |
| Concentration | 1.04 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 15mM sodium azide; pH 7.4 |
| Gene | FN1 |
| Gene Accession No. | P02751, P07589, P11722 |
| Gene Alias | Anastellin; CIG; Cold insoluble globulin (CIG); Cold-insoluble globulin; cumulus cell-specific fibronectin 1 transcript; DKFZp686F10164; DKFZp686H0342; DKFZp686I1370; DKFZp686O13149; E330027I09; ED-B; embryo-specific fibronectin 1 transcript; ferritin L subunit; ferritin L-chain; FIBNEC; fibronectin; fibronectin 1; fibronectin ED-A- region; fibronectin ED-B+ region; fibronectin variable region; FINC; FN; FN1; fn-1; FNZ; GFND; GFND2; LETS; LOW QUALITY PROTEIN: fibronectin; Migration stimulating factor (MSF); migration-stimulating factor; MSF; Ugl-Y1; Ugl-Y2; Ugl-Y3 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.