missing translation for 'onlineSavingsMsg'
Learn More

Ficoll 400 (White to Off-White Powder/Molecular Biology), Fisher BioReagents™

Product Code. p-25894
Change view
Click to view available options
Quantity:
10 g
25 g
100 g
500 g
Packaging:
Poly Bottle
Unit Size:
100g
10g
25g
500g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10295003 10 g Poly Bottle 10g
10071593 25 g Poly Bottle 25g
10468343 100 g Poly Bottle 100g
10478343 500 g Poly Bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10295003 Supplier Fisher BioReagents Supplier No. BP52510
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Ficoll 400 (White to Off-White Powder/Molecular Biology)

Ficoll 400 is used as a component of Denhardt's Reagent and in gel-loading buffers for DNA gels. It is also used in forming density gradients to separate cells and subcellular components.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 26873-85-8
Molecular Formula C15H27ClO12
Molecular Weight (g/mol) 434.819
InChI Key IACFXVUNKCXYJM-AKSHDPDZSA-N
Synonym ficol 400
PubChem CID 88330506
IUPAC Name 2-(chloromethyl)oxirane;(2R,3R,4S,5S,6R)-2-[(2S,3S,4S,5R)-3,4-dihydroxy-2,5-bis(hydroxymethyl)oxolan-2-yl]oxy-6-(hydroxymethyl)oxane-3,4,5-triol
SMILES C1C(O1)CCl.C(C1C(C(C(C(O1)OC2(C(C(C(O2)CO)O)O)CO)O)O)O)O

Specifications

Color White
Physical Form Powder/Solid
Quantity 10 g
Formula Weight ≈400,000
Grade Molecular Biology
Packaging Poly Bottle
Specific Rotation Inclusive between 50° (+ or -)
Chemical Name or Material Ficoll™ 400
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.