accessibility menu, dialog, popup

UserName

OClOOOOHOHOHOHOHHOHOOH

CAS RN 26873-85-8

IUPAC Name: 2-(chloromethyl)oxirane; 2-{[3,4-dihydroxy-2,5-bis(hydroxymethyl)oxolan-2-yl]oxy}-6-(hydroxymethyl)oxane-3,4,5-triol
Synonyms: epichlorohydrin; granulated sugar
Molecular Weight (g/mol): 434.82
Molecular Formula: C15H27ClO12
InChi Key: IACFXVUNKCXYJM-UHFFFAOYNA-N
SMILES: ClCC1CO1.OCC1OC(CO)(OC2OC(CO)C(O)C(O)C2O)C(O)C1O
Quantity 
  • (4)
  • (4)
  • (1)
  • (4)
  • (2)
  • (1)
  • (1)
  • (1)
Grade 
  • (6)
pH 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (3)
  • (7)
  • (2)
  • (2)
  • (4)
  • (4)
  • (1)
  • (4)
  • (2)
  • (1)
  • (1)
  • (1)
  • (6)
  • (2)
  • (2)