missing translation for 'onlineSavingsMsg'
Learn More

FICOLL™, White Powder, MP Biomedicals™

Product Code. 11480182 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
10 g
100 g
Unit Size:
100g
10g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11480182 100 g 100g
11460182 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11480182 Supplier MP Biomedicals Supplier No. 0216000380
 Restricted Item
Estimated Shipment: 15-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 26873-85-8 | C15H27ClO12 | 434.819 g/mol

Chemical Identifiers

CAS 26873-85-8
Molecular Formula C15H27ClO12
Molecular Weight (g/mol) 434.819
InChI Key IACFXVUNKCXYJM-AKSHDPDZSA-N
Synonym ficol 400
PubChem CID 88330506
IUPAC Name 2-(chloromethyl)oxirane;(2R,3R,4S,5S,6R)-2-[(2S,3S,4S,5R)-3,4-dihydroxy-2,5-bis(hydroxymethyl)oxolan-2-yl]oxy-6-(hydroxymethyl)oxane-3,4,5-triol
SMILES C1C(O1)CCl.C(C1C(C(C(C(O1)OC2(C(C(C(O2)CO)O)O)CO)O)O)O)O

Specifications

Color White
Density up to 1.2g/mL
Quantity 100 g
Formula Weight ≈400,000
Physical Form Solid
Chemical Name or Material FICOLL™
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.