missing translation for 'onlineSavingsMsg'
Learn More

Fingolimod hydrochloride, MedChemExpress

Product Code. 16477657
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
200 mg
1 g
5 g
Unit Size:
100mg
1g
200mg
5g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16477657 5 g 5g
16451219 1 g 1g
16445686 100 mg 100mg
16411026 200 mg 200mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16477657 Supplier Medchem Express Supplier No. HY12005
 Restricted Item
Estimated Shipment: 07-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Fingolimod hydrochloride (FTY720), an analog of sphingosine, is a potent sphingosine 1-phosphate (S1P) receptors modulator. Fingolimod hydrochloride is phosphorylated by sphingosine kinases, particularly by SK2, and then binds S1PR1, 3, 4, and 5. Fingolimod hydrochloride induces the internalization of S1P1, and consequently, inhibits S1P activity. Fingolimod hydrochloride also is a pak1 activator.

Fingolimod hydrochloride (FTY720), an analog of sphingosine, is a potent sphingosine 1-phosphate (S1P) receptors modulator. Fingolimod hydrochloride is phosphorylated by sphingosine kinases, particularly by SK2, and then binds S1PR1, 3, 4, and 5. Fingolimod hydrochloride induces the internalization of S1P1, and consequently, inhibits S1P activity. Fingolimod hydrochloride also is a pak1 activator.

Specifications

Chemical Name or Material Fingolimod hydrochloride
Physical Form Solid
CAS 162359-56-0
Color White
Quantity 5 g
Purity Grade Notes Research
Molecular Formula C19H34ClNO2
For Use With (Application) COVID-19-anti-virus
Synonym FTY720
Solubility Information DMSO : ≥ 100 mg/mL (290.76 mM) ∣H2O : 50 mg/mL (145.38 mM; Need ultrasonic)
SMILES OCC(N)(CO)CCC1=CC=C(C=C1)CCCCCCCC.[H]Cl
Molecular Weight (g/mol) 343.93
Formula Weight 343.93
Percent Purity 95.0%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.