missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisher BioReagents™ 2M Calcium Chloride, Microbial Cell Culture Grade
Description
- CAS: 10035-04-08 (Calcium Chloride Dihydrate), 7732-18-5 (Water)
- Concentration: 294g/L
- Calcium chloride solution is commonly used in cell biology to prepare competent E. coli cells with improved transformation efficiency
- Calcium chloride increases the ability of the cell to incorporate plasmid DNA due to the postitively charged calcium ions
- Promotes the binding of plasmid DNA to lipopolysaccharides
- Product is supplied as 5 x 10mL
- Intended for research use only
Specifications
Specifications
| Concentration | 294g/L |
| Form | Liquid |
| Grade | Microbial cell culture |
| Manufacturing Origin | United States |
| Fill Volume | 50 mL |
| For Use With (Application) | Widely used to prepare competent Escherichia coli strains |
| Packaging Type | Bottle |
| Quantity | 50 mL |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?