missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisher BioReagents™ 2M Magnesium Chloride, Microbial Cell Culture Grade
Description
- CAS: 7791-18-6 (Magnesium Chloride Hexahydrate), 7732-18-5 (Water)
- Concentration: 406.6g/L
Specifications
Specifications
| Concentration | 406.6g/L |
| Form | Liquid |
| Grade | Microbial cell culture |
| Manufacturing Origin | United States |
| Media Type | Magnesium Chloride |
| Fill Volume | 10 mL |
| For Use With (Application) | For addition to SOB and SOC medium for propagating bacterial strains |
| Packaging Type | Bottle |
| Quantity | 10 mL |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?