Learn More
Fisherbrand™ Stackable Microplate Holder for Microplate Vortex Mixers
For use with Microplate Vortex Mixers.02-216-108 and 02-216-109
Brand: Fisherbrand™
Additional Details : Weight : 0.16000kg
Description
- To maximize capacity of Heavy-Duty Vortex Mixer to eight microplates by stacking the tray on top of four plate holder
Specifications
Stackable microplate holder | |
Microplate vortex mixers 02-216-108 and 02-216-109 | |
8 |