missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Vortex Mixer Accessories Kits

Product Code. 15726949 Shop All Fisherbrand Traceable Products
Change view
Click to view available options
Product Type:
Foam Microtube and Microwell Plate Kit w/ Retainer
Foam Tube Holder Kit w/ Retainer
Microtube Kit
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Product Type Unit Size
15726949 Foam Microtube and Microwell Plate Kit w/ Retainer Each
15756939 Foam Tube Holder Kit w/ Retainer Each
15766959 Microtube Kit Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 15726949
  • ONLINE EXCLUSIVE PRICE
    96.75 €
    69.09 € / Pack of 1
    Save 27.66 €
    28% Off
Estimated Shipment: 16-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

For use with applicable Fisherbrand vortex mixers.

Foam Microtube and Microwell Plate Holder Kit with Retainer

  • Includes one microtube holder, one microwell plate holder, and one insert retainer (Cat. Nos. 02-215-386, -387, -391)

Foam Tube Holder Kit with Retainer

  • Iincludes 3 tube holders (for 9-13mm, 14-19mm, and 20-25mm tubes), one flat foam insert, two vessel harnesses and one insert retainer (Cat. Nos. 02-215-393, -394, -395, -397, -392, -391)
Microtube Kit:
  • Mixes (48) 0.25 to 2mL micro tubes; includes foam insert and insert retainer
Microwell Plate Kit:
  • For mixing 96-well plates or deep well blocks; includes foam insert and insert retainer

 

 

TRUSTED_SUSTAINABILITY

Specifications

Holds 0.25 to 2 mL
Product Type Foam Microtube and Microwell Plate Kit w/ Retainer
For Use With (Equipment) Vortex mixer
Includes One microtube holder, one microwell plate holder, and one insert retainer
No. of Positions 48
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.