missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ 1536-Well Polystyrene Plates

Product Code. 13575440 Shop All Fisherbrand Products
Change view
Click to view available options
Color:
Black
Clear
White
Unit Size:
Pack of 100
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color unitSize
13575440 Clear Pack of 100
13595440 White Pack of 100
13585440 Black Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 13575440

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternative produkter

Engineered for high throughput screening, allowing end users to screen four times as many samples in one plates

Optimized for robotic instruments, plate readers and liquid handling systems

  • Highly specified polystyrene
  • Available in black, white and clear
  • Low plate height optimized for stacking efficiency
  • Working volume range: 1–10lμL/well
  • Untreated
  • Non-sterile

Ordreinfo

Lids sold separately

TRUSTED_SUSTAINABILITY

Tekniske data

Format Standard
Lid Without Lid
Array 32 x 48
Color Clear
Product Type 1536-Well Cell Culture Multiwell Plates
Material Polystyrene
No. of Wells 1536
Volume (Metric) Working 1 to 101 μL/well
Sterility Non-sterile
Surface Treatment Non-treated
Well Shape Flat Bottom (F-well)
For Use With (Application) For low volume assays such as colorimetric, enzymatic, cloning, replication and array, as well as storage
Vis mere Vis mindre
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.