missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ 1536-Well Polystyrene Plates

Product Code. 13595440 Shop All Fisherbrand Products
Change view
Click to view available options
Color:
Black
Clear
White
Unit Size:
Pack of 100
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color unitSize
13595440 White Pack of 100
13575440 Clear Pack of 100
13585440 Black Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 13595440

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

Engineered for high throughput screening, allowing end users to screen four times as many samples in one plates

Optimized for robotic instruments, plate readers and liquid handling systems

  • Highly specified polystyrene
  • Available in black, white and clear
  • Low plate height optimized for stacking efficiency
  • Working volume range: 1–10lμL/well
  • Untreated
  • Non-sterile

Order Info

Lids sold separately

TRUSTED_SUSTAINABILITY

Specifications

Format Standard
Lid Without Lid
Array 32 x 48
Color White
Product Type 1536-Well Cell Culture Multiwell Plates
Material Polystyrene
No. of Wells 1536
Volume (Metric) Working 1 to 101 μL/well
Sterility Non-sterile
Surface Treatment Non-treated
Well Shape Flat Bottom (F-well)
For Use With (Application) For low volume assays such as colorimetric, enzymatic, cloning, replication and array, as well as storage
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.