missing translation for 'onlineSavingsMsg'
Learn More

Traceable™ One-Shot Conductivity Standards, Fisherbrand™

Product Code. 11724256 Shop All Fisherbrand Products
Change view
Click to view available options
Concentration:
150,000 μS/μ℧
200,000 μS/μ℧
Unit Size:
Pack of 6
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Concentration unitSize
11724256 150,000 μS/μ℧ Pack of 6
11734256 200,000 μS/μ℧ Pack of 6
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11724256
  • ONLINE EXCLUSIVE PRICE
    51.90 €
    47.44 € / Pack of 6
    Save 4.46 €
    8% Off
 Restricted Item
Estimated Shipment: 02-10-2026
Add to basket
Add to basket

Eliminates contamination with calibration within disposable container. Traceable™ One-Shot Conductivity Standards are 100% compatible with all conductivity meters.

  • Nominal Value: 150000 Micro (mhos/siemens)
  • Certified Reference Material
  • Accuracy: certificate stated uncertainty, or ±0.25 microsiemen or ±0.25%
  • Includes: 6-pack 100 mL container, step-by-step calibration instructions, individual temperature compensation charts, traceability information, Traceable Certificate

Specifications

Chemical Name or Material One-Shot Conductivity Standards
Packaging Bottle
CAS 7447-40-7
Molecular Formula ClK
Concentration 150,000 μS/μ℧
MDL Number MFCD00011360
Technique Calibration of Conductivity Instrument
Color Clear
For Use With (Equipment) Conductivity Meters and Probes
Solubility Soluble in water
Solution Type Calibration Solution
InChI Key WCUXLLCKKVVCTQ-UHFFFAOYSA-M
Type Conductivity Standard
SMILES [Cl-].[K+]
IUPAC Name potassium chloride
Molecular Weight (g/mol) 74.548
PubChem CID 4873
Quantity 6 x 100 mL
ChEBI CHEBI:32588
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.