missing translation for 'onlineSavingsMsg'
Learn More

FKBP10/FKBP65 Rabbit Recombinant Antibody, Proteintech

Product Code. 30235543 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
100 μL
20 μL
Unit Size:
100µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30235543 20 μL 20µL
30237186 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30235543 Supplier Proteintech Supplier No. 815671RR20UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Recombinant Antibody

The FKBP10/FKBP65 antibody from Proteintech is a rabbit recombinant antibody generated with recombinant protein of human FKBP10/FKBP65. This antibody recognizes human antigen. The FKBP10/FKBP65 antibody has been validated for the following applications: WB, IHC, ELISA analysis.

Specifications

Antigen FKBP10/FKBP65
Applications Western Blot, Immunohistochemistry, ELISA
Classification Recombinant Monoclonal
Clone S94
Concentration 500 μg/mL
Conjugate Unconjugated
Gene Symbols FKBP6; FKBP65; FLJ20683; FLJ22041; FLJ23833; hFKBP65
Host Species Rabbit
Immunogen Recombinant Protein
Purification Method Protein A purification
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 60681
Target Species Human
Content And Storage Store at -20°C. Stable for one year after shipment.
Antibody Molecular Weight 65-70 kDa
Product Type Recombinant Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.