missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FKBP52/FKBP4 Antibody (S08-4F8), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-19864-50ul
This item is not returnable.
View return policy
Description
FKBP52/FKBP4 Monoclonal antibody specifically detects FKBP52/FKBP4 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| FKBP52/FKBP4 | |
| Monoclonal | |
| 0.3 mg/mL | |
| Western Blot, Immunohistochemistry 1:200, Immunohistochemistry-Paraffin | |
| 51 kDa FK506-binding protein, 52 kDa FKBP, 59 kDa immunophilin, EC 5.2.1.8, FK506 binding protein 4, 59kDa, FK506-binding protein 4, FK506-binding protein 4 (59kD), FKBP-4, FKBP51, FKBP-52, FKBP52T-cell FK506-binding protein, 59kD, FKBP5952 kDa FK506-binding protein, HBI, HSP binding immunophilin, Hsp56, HSP-binding immunophilin, Immunophilin FKBP52, p52, p59, peptidylprolyl cis-trans isomerase, peptidyl-prolyl cis-trans isomerase FKBP4, PPIase, PPIase FKBP4, Rotamase | |
| A synthetic peptide of human FKBP52/FKBP4 (Uniprot # Q02790) | |
| 50 μg | |
| Membrane Trafficking and Chaperones, Stem Cell Markers | |
| 2288 | |
| Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| S08-4F8 | |
| Unconjugated | |
| 50mM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction