Learn More
Novus Biologicals™ FLAP Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand:  Novus Biologicals™ NBP3-30926-100ug
 View more versions of this product
Product Code. 30066911
  • 508.00 € / 100µg
Description
FLAP Polyclonal antibody specifically detects FLAP in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY
Specifications
Specifications
FLAP
Polyclonal
Western Blot : 1-5 μg/mL
arachidonate 5-lipoxygenase-activating protein, FLAPMK-886-binding protein
Synthetic peptide derived from human FLAP protein.
100 μg
Immunology, Inflammation
241
Store at -20°C. Avoid freeze/thaw cycles.
Western Blot
Unconjugated
Phosphate buffered saline
Rabbit
Ammonium sulfate precipitation
RUO
Primary
Human
Purified