missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Flavivirus group antigen Antibody (D1-4G2-4-15 (4G2)) - Azide and BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
259.00 € - 589.00 €
Specifications
| Antigen | Flavivirus group antigen |
|---|---|
| Clone | D1-4G2-4-15 (4G2) |
| Dilution | Western Blot 1:100 - 1:2000, Flow Cytometry 1:10 - 1:1000, ELISA 1:100 - 1:2000, Immunohistochemistry-Paraffin, Immunofluorescence 1:10 - 1:500, CyTOF-ready |
| Applications | Western Blot, Flow Cytometry, ELISA, Immunohistochemistry (Paraffin), Immunofluorescence, CyTOF |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18287810
|
Novus Biologicals
NBP2-52709-0.025MG |
0.025 mg |
259.00 €
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18073876
|
Novus Biologicals
NBP2-52709-0.2MG |
0.2 mg |
589.00 €
0.20mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Flavivirus group antigen Monoclonal specifically detects Flavivirus group antigen in Monkey, Virus samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Functional, Immunofluorescence, Binding Activity, CyTOF-ready.Specifications
| Flavivirus group antigen | |
| Western Blot 1:100 - 1:2000, Flow Cytometry 1:10 - 1:1000, ELISA 1:100 - 1:2000, Immunohistochemistry-Paraffin, Immunofluorescence 1:10 - 1:500, CyTOF-ready | |
| Monoclonal | |
| Purified | |
| RUO | |
| PBS with 0.02% Proclin 300. with 0.02% Proclin 300 | |
| Dengue Virus type 2 antigens. | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| D1-4G2-4-15 (4G2) | |
| Western Blot, Flow Cytometry, ELISA, Immunohistochemistry (Paraffin), Immunofluorescence, CyTOF | |
| Unconjugated | |
| Mouse | |
| Virus | |
| DEN 1, DEN1, Dengue hemorrhagic fever, Dengue type 1 virus, DENV 1, DENV1 | |
| IgG2a | |
| Protein A or G purified | |
| Recognises flavivirus group specific antigens (Dengue virus, West Nile Virus, Japanese Encephalitis, Zika virus etc). It binds to the fusion loop at the extremity of domain II of protein E. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title