Learn More
Invitrogen™ Fletikumab Recombinant Monoclonal Antibody
Recombinant Monoclonal Antibody
Brand:  Invitrogen™ MA558651
 View more versions of this product
Product Code. 30283359
  • 477.00 € / 100µg
Description
For reconstitution, add sterile, distilled water to achieve a final antibody concentration of 1 mg/mL. Gently shake to solubilize the protein completely. Do not vortex. Reconstituted products should be stored at -80 °.
Fletikumab is under investigation in clinical trial NCT01038674 (Safety and Tolerability of Anti-IL-20 in Subjects With Rheumatoid Arthritis).
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Fletikumab
Recombinant Monoclonal
Unconjugated
15D2; NN-8226; NNC-0109-0012
Protein A
RUO
Human
Antibody
IgG4SP
ELISA, Flow Cytometry, Functional Assay
1 mg/mL
25mM histidine with 8% sucrose, 0.01% Tween 80 and no preservative; pH 6.2
Human
100 μg
Primary
-20°C, Avoid Freeze/Thaw Cycles
Lyophilized